Learn More
RAPH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554127
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NPQPQQWSKM SVKKAPPPTR PKRNDSTRLT QAEISEQPTM ATVVPQVPTS PKSSLSVQPG FLADLNRTLQ RKSITRHGSL SSRMSRAEPT ATMDDMALPP PPPELLSDQQ KAGYGGSHIS GYATLRRGPP PAPPKRDQNT KLSR Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
RAPH1 Polyclonal antibody specifically detects RAPH1 in Human samples. It is validated for Immunohistochemistry.Specifications
RAPH1 | |
Polyclonal | |
Unconjugated | |
RAPH1 | |
9430025M21Rik; ALS2CR18; ALS2CR9; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 18; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 9; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 18 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 9 protein; C730009O10Rik; KIAA1681; Lamellipodin; LPD; PREL2; PREL-2; proline-rich EVH1 ligand 2; Protein RMO1; RalGDS/AF-6; RAPH1; Ras association (RalGDS/AF-6) and pleckstrin homology domains 1; ras-associated and pleckstrin homology domains-containing protein 1; RMO1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65059 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q70E73 | |
RAPH1 | |
Recombinant protein corresponding to Human RAPH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only