missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Range Finder Hydrometer

Brand:  Brand™ 970010

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10528031

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

1Pack
0.700 to 1.000g/cm3
Range Finder Hydrometer
260 to 300mm
0.005g/cm3
Videos