missing translation for 'onlineSavingsMsg'
Learn More

Raltegravir potassium salt, Thermo Scientific Chemicals

Catalog Number p-7112905
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871038-72-1
C20H20FKN6O5
482.51
IFUKBHBISRAZTF-UHFFFAOYSA-M
raltegravir potassium, raltegravir potassium salt, raltegravirpotassiumsalt, raltegravir mk-0518, potassium 4-4-fluorobenzyl carbamoyl-1-methyl-2-2-5-methyl-1,3,4-oxadiazole-2-carboxamido propan-2-yl-6-oxo-1,6-dihydropyrimidin-5-olate, isentress tn, raltegravir potassium usan:jan, pubchem22484, n-1-4-4-fluorophenyl methylcarbamoyl-5-hydroxy-1-methyl-6-oxo-pyrimidin-2-yl-1-methyl-ethyl-5-methyl-1,3,4-oxadiazole-2-carboxamide
23668479
potassium;4-[(4-fluorophenyl)methylcarbamoyl]-1-methyl-2-[2-[(5-methyl-1,3,4-oxadiazole-2-carbonyl)amino]propan-2-yl]-6-oxopyrimidin-5-olate
CC1=NN=C(O1)C(=O)NC(C)(C)C2=NC(=C(C(=O)N2C)[O-])C(=O)NCC3=CC=C(C=C3)F.[K+]
This item is not returnable. View return policy
871038-72-1
C20H20FKN6O5
482.51
IFUKBHBISRAZTF-UHFFFAOYSA-M
raltegravir potassium, raltegravir potassium salt, raltegravirpotassiumsalt, raltegravir mk-0518, potassium 4-4-fluorobenzyl carbamoyl-1-methyl-2-2-5-methyl-1,3,4-oxadiazole-2-carboxamido propan-2-yl-6-oxo-1,6-dihydropyrimidin-5-olate, isentress tn, raltegravir potassium usan:jan, pubchem22484, n-1-4-4-fluorophenyl methylcarbamoyl-5-hydroxy-1-methyl-6-oxo-pyrimidin-2-yl-1-methyl-ethyl-5-methyl-1,3,4-oxadiazole-2-carboxamide
23668479
potassium;4-[(4-fluorophenyl)methylcarbamoyl]-1-methyl-2-[2-[(5-methyl-1,3,4-oxadiazole-2-carbonyl)amino]propan-2-yl]-6-oxopyrimidin-5-olate
CC1=NN=C(O1)C(=O)NC(C)(C)C2=NC(=C(C(=O)N2C)[O-])C(=O)NCC3=CC=C(C=C3)F.[K+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155.0°C to 157.0°C
Quantity 250 mg
Formula Weight 482.51
Chemical Name or Material Raltegravir potassium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'