missing translation for 'onlineSavingsMsg'
Learn More

Ralfinamide mesylate, Tocris Bioscience™

Na+ channel blocker

Brand:  Tocris (Bio-Techne) 402950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443984

  • £462.82 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sodium channel blocker. Suppresses tetrodotoxin (TTX)-resistant Na+ currents approximately twice as selectively as TTX-sensitive currents. Antinociceptive; displays analgesic effects in animal models of inflammatory and neuropathic pain.
Specifications

Chemical Identifiers

202825-45-4
398.449
ralfinamide mesylate, s-2-4-2-fluorobenzyl oxy benzyl amino propanamide methanesulfonate, 2s-2-4-2-fluorophenyl methoxy phenyl methyl amino-propanamide methanesulfonate, methanesulfonic acid; ralfinamide, ralfinamide mesylate hplc, s-2-4-2-fluorobenzyl oxy benzyl-amino propanamide methanesulfonate, s-2-4-2-fluorobenzyloxy benzylamino propanamide methanesulfonate, 2s-2-4-2-fluorophenyl methoxy phenyl methylamino propanamide; methanesulfonic acid, methanesulfonic acid-n~2~-4-2-fluorophenyl methoxy phenyl methyl-l-alaninamide 1/1
(2S)-2-[[4-[(2-fluorophenyl)methoxy]phenyl]methylamino]propanamide;methanesulfonic acid
C18H23FN2O5S
CHQVNINIGBRKGZ-YDALLXLXSA-N
23661379
CC(C(=O)N)NCC1=CC=C(C=C1)OCC2=CC=CC=C2F.CS(=O)(=O)O

Specifications

Ralfinamide mesylate
202825-45-4
ralfinamide mesylate, s-2-4-2-fluorobenzyl oxy benzyl amino propanamide methanesulfonate, 2s-2-4-2-fluorophenyl methoxy phenyl methyl amino-propanamide methanesulfonate, methanesulfonic acid; ralfinamide, ralfinamide mesylate hplc, s-2-4-2-fluorobenzyl oxy benzyl-amino propanamide methanesulfonate, s-2-4-2-fluorobenzyloxy benzylamino propanamide methanesulfonate, 2s-2-4-2-fluorophenyl methoxy phenyl methylamino propanamide; methanesulfonic acid, methanesulfonic acid-n~2~-4-2-fluorophenyl methoxy phenyl methyl-l-alaninamide 1/1
CC(C(=O)N)NCC1=CC=C(C=C1)OCC2=CC=CC=C2F.CS(=O)(=O)O
398.449
398.45
50mg
C18H23FN2O5S
CHQVNINIGBRKGZ-YDALLXLXSA-N
(2S)-2-[[4-[(2-fluorophenyl)methoxy]phenyl]methylamino]propanamide;methanesulfonic acid
23661379
>99%
Videos