Learn More
Mettler Toledo™ Rainin™ BioClean™ Bulk Pipette Tips
BioClean™ tips provided in economical bulk bags for use with Rainin™ pipettes with LTS™ LiteTouch™.
Brand:  Mettler Toledo™ 17001119
Code : Z2
Additional Details : Weight : 0.95850kg
 View more versions of this product
Product Code. 12150681
  • £90.45 / Pack of 200
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Flawless performance: perfect production and advanced design
  • No deformations, cracks or flashes
  • 100% inert: purity and cleanliness are assured
  • All tips are 100% free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors and additives
  • Packaged in a re-sealable zip-lock bag, for manual rack refilling
Specifications
Specifications
1000 μL
Non-sterile
Rainin™ Pipette with LTS™ LiteTouch™
Non-Filtered
1000 tips/bag
Rainin BioClean