Learn More
RAE1 delta Monoclonal Antibody (RD-41), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-5756-81
38.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The RD-41 monoclonal antibody reacts with the murine Rae1delta. Rae-1delta is one of several known murine NKG2D ligands that include RAE-1 molecules (alpha, beta, ε, gamma and delta), H60 and MULT-1. Expression of NKG2D ligands is low or absent on normal adult tissues. However, stressed or transformed cells express NKG2D ligands which in turn activates NK cells tumoricidal activity through NKG2D. Until now, the expression of NKG2D ligands has been mainly studied with NKG2D tetramers which recognizes all NKG2D ligands. The RD-41 antibody has been reported to block tetramer staining. Applications Reported: This RD-41 antibody has been reported for use in flow cytometric analysis, immunohistology staining of frozen tissue sections and ELISA. Applications Tested: This RD-41 antibody has been tested by flow cytometric analysis of dendritic cell line. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Rae-1 delta is one of several known murine NKG2D ligands that include RAE-1 molecules, H60 and MULT-1. Expression of NKG2D ligands is low or absent on normal adult tissues. However, stressed or transformed cells express NKG2D ligands which in turn activates NK cells tumoricidal activity through NKG2D. Until now, the expression of NKG2D ligands has been mainly studied with NKG2D tetramers which recognizes all NKG2D ligands.Specifications
RAE1 delta | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9JI58 | |
Raet1d | |
Affinity chromatography | |
RUO | |
56554 | |
4° C | |
Liquid |
ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
RD-41 | |
Unconjugated | |
Raet1d | |
RAE-1delta; RAE-1-delta; Raet1d; retinoic acid early transcript 1D; retinoic acid early transcript delta; retinoic acid early-inducible protein 1-delta | |
Mouse | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
For Research Use Only.