missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Rae-1 alpha/beta/delta/epsilon Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | Rae-1 |
---|---|
Clone | 199205 |
Concentration | LYOPH |
Dilution | Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
Applications | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Description
Rae-1 Monoclonal specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
Rae-1 | |
LYOPH | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
8480 | |
BaF3 mouse pro-B cell line transfected with Rae-1 beta (Accession # NP_033043) and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 (Accession # Q9JI58) | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Rae-1 alpha/beta/delta/epsilon . When tested in flow cytometry against a panel of transfectants expressing different Rae-1 proteins, this antibody strongly recognizes Rae-1 beta and Rae-1 delta . It shows weaker staining of Rae-1 alpha and Rae-1 epsilon transfectants. It does not recognize Rae-1 gamma . |
199205 | |
Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Raet1a | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only