missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Radiation Hazard Trays
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
£74.50 - £81.45
Specifications
Color | Yellow |
---|---|
Material | APET |
Type | Radiation Hazard Tray |
For Use With | Lab bench |
Shape | Rectangular |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12317247
|
Fisherbrand™
|
54 x 34 cm |
£74.50
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
12347247
|
Fisherbrand™
|
70 x 46 cm |
£81.45
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
- Radiation hazard tray yellow
- Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Yellow | |
Radiation Hazard Tray | |
Rectangular |
APET | |
Lab bench |