missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550025
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total RAD18 protein.
RAD18 Polyclonal antibody specifically detects RAD18 in Human samples. It is validated for Western Blot.Specifications
RAD18 | |
Polyclonal | |
Unconjugated | |
Rad18 | |
2810024C04Rik; E3 ubiquitin-protein ligase RAD18; hHR18; hRAD18; mRAD18Sc; postreplication repair protein hRAD18p; postreplication repair protein RAD18; post-replication repair protein RAD18SC; Rad18; RAD18 E3 ubiquitin protein ligase; RAD18 homolog; RAD18 homolog (S. cerevisiae); RAD18, E3 ubiquitin protein ligase; RAD18, S. cerevisiae, homolog; Rad18sc; RING finger protein 73; RING-type E3 ubiquitin transferase RAD18; RNF73 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56852 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NS91 | |
Rad18 | |
Synthesized peptide derived from internal of human RAD18. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |