missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD18 Antibody (79B1048.1) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56523JF669
This item is not returnable.
View return policy
Description
RAD18 Monoclonal antibody specifically detects RAD18 in Human samples. It is validated for Western Blot
Specifications
| RAD18 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 6.3.2.-, hHR18, hRAD18, postreplication repair protein hRAD18p, Postreplication repair protein RAD18, RAD18 homolog (S. cerevisiae), RAD18, S. cerevisiae, homolog, RING finger protein 73, RNF73E3 ubiquitin-protein ligase RAD18 | |
| The antibody was raised against a synthetic peptide corresponding to the amino acids 402-414 of human Rad18. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers, Zinc Finger | |
| 56852 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| 79B1048.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction