Learn More
Hamilton™ Non-Filtered Racked Pipette Tips
Delivering outstanding accuracy and precision, guaranteed.
£41.15 - £82.35
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | ||||
15365084
|
Hamilton™
68009-11 |
300 μL | Non-sterile |
£67.00
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
15395094
|
Hamilton™
68007-11 |
10 μL | Non-sterile |
£67.85
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
15305084
|
Hamilton™
68010-12 |
1000 μL | Sterile |
£41.15
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
15305094
|
Hamilton™
68008-12 |
200 μL | Sterile |
£68.65
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
15385094
|
Hamilton™
68007-12 |
10 μL | Sterile |
£82.35
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Pre-sterilized or non-sterile
- 96 tips to a rack
Specifications
Non-Filtered |