missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Classic Rack for Chest Freezers, 100 mm Box Height, with Boxes

Classic Rack is suitable for use with all chest freezer models.

Brand:  Tenak™ TE21472B

Additional Details : Weight : 1.80000kg

 View more versions of this product

Product Code. 15280504

  • £94.30 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Comes with a locking rod to secure the cryoboxes
  • Measure the inside height of you freezer when choosing the right rack
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

White cardboard boxes, 9 x 9 cell dividers
Stainless Steel
140 x 140 x 530 mm
Boxes (5)
Chest Frseezers
100 mm (Box)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers