missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 16 mm Shelf Height

Eco AluRack for chest freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE15110

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12418170

  • £119.00 / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket

Description

Description

  • For use in chest freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications

Specifications

Microtiterplates (32)
Chest Frseezers
16 mm
Aluminum
135 x 92 x 555 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers