missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RACGAP1 Mouse anti-Human, Mouse, Rat, Clone: 6E10A2, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | RACGAP1 |
---|---|
Clone | 6E10A2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877585
|
Proteintech
66056-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867585
|
Proteintech
66056-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rho GTPases control a variety of cellular processes. There are 3 subtypes of Rho GTPases in the Ras superfamily of small G proteins: RHO (see MIM 165370), RAC (see RAC1; MIM 602048), and CDC42 (MIM 116952). GTPase-activating proteins (GAPs) bind activated forms of Rho GTPases and stimulate GTP hydrolysis. Through this catalytic function, Rho GAPs negatively regulate Rho-mediated signals. GAPs may also serve as effector molecules and play a role in signaling downstream of Rho and other Ras-like GTPases.Specifications
RACGAP1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HsCYK 4, ID GAP, KIAA1478, Male germ cell RacGap, MgcRacGAP, RACGAP1 | |
Racgap1 | |
IgG1 | |
Protein G | |
Antibody |
6E10A2 | |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q9H0H5, Q9WVM1 | |
26934, 29127, 315298 | |
RACGAP1 Fusion Protein Ag6944 | |
Primary | |
-20°C | |
RACGAP1 |