missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ RAC1 Recombinant Human Protein, N-GST Tag

Expressed from DNA sequence encoding mature form of human Rac1 isoform A (Met 1-Cys 189) fused to GST tag at N-terminus

Brand:  Sino Biological™ 10535H09B5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15236895

  • £141.58 / 5µg


Estimated Shipment
08-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 50mM Tris, 100mM NaCl, 1mM GSH, 10% Glycerol, pH 8.0.
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
5879
≥90% by SDS-PAGE
Human
<1 EU/μg
RAC 1
Other proteins
RAC1 Recombinant Human Protein
Lyophilized
47kd
5μg
Full Length, Recombinant
Ras-Related C3 Botulinum Toxin Substrate 1 Protein
Assay Standard
Insect
N-GST Tag
Videos