Learn More
Sino Biological™ RAC1 Recombinant Human Protein, N-GST Tag
Expressed from DNA sequence encoding mature form of human Rac1 isoform A (Met 1-Cys 189) fused to GST tag at N-terminus
Brand: Sino Biological™ 10535H09B50
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Expressed from DNA sequence encoding mature form of human Rac1 isoform A (Met 1-Cys 189) fused to GST tag at N-terminus
- Formulation: 50mM Tris, 100mM NaCl, 1mM GSH, 10% Glycerol, pH 8.0
Specifications
Protein Structure Analysis, Immunogen, Protein Standard, Cell Biology and Analysis | |
47kd | |
Human | |
<1 EU/μg | |
RAC 1 | |
N-GST Tag | |
Other proteins | |
≥90% by SDS-PAGE |
5879 | |
50μg | |
Full Length, Recombinant | |
Ras-Related C3 Botulinum Toxin Substrate 1 Protein | |
RAC1 Recombinant Human Protein | |
Insect | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.