missing translation for 'onlineSavingsMsg'
Learn More

rac BHFF, Tocris Bioscience™

Potent and selective postitive allosteric modulator of GABAB receptors

Brand:  Tocris (Bio-Techne) 331310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13452764

  • £163.17 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective GABAB receptor positive allosteric modulator that increases the potency and efficacy of GABA ( > 15-fold and > 149% respectively). Exhibits anxiolytic activity in vivo and is orally active.
Specifications

Chemical Identifiers

123557-91-5
330.347
rac bhff, bhff, 2 3h-benzofuranone,5,7-bis 1,1-dimethylethyl-3-hydroxy-3-trifluoromethyl, acmc-20emwz, rac bhff hplc, 5,7-ditert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2-one, 3h-benzofuran-2-one, 5,7-di-tert-butyl-3-hydroxy-3-trifluoromethyl, 5,7-di tert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2 3h-one, 5,7-di-tert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2 3h-one, 5,7-bis 1,1-dimethylethyl-3-hydroxy-3 trifluoromethyl-2 3h-benzofuranone
5,7-ditert-butyl-3-hydroxy-3-(trifluoromethyl)-1-benzofuran-2-one
C17H21F3O3
RVNOANDLZIIFHB-UHFFFAOYSA-N
4332683
CC(C)(C)C1=CC(=C2C(=C1)C(C(=O)O2)(C(F)(F)F)O)C(C)(C)C

Specifications

10mg
123557-91-5
rac bhff, bhff, 2 3h-benzofuranone,5,7-bis 1,1-dimethylethyl-3-hydroxy-3-trifluoromethyl, acmc-20emwz, rac bhff hplc, 5,7-ditert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2-one, 3h-benzofuran-2-one, 5,7-di-tert-butyl-3-hydroxy-3-trifluoromethyl, 5,7-di tert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2 3h-one, 5,7-di-tert-butyl-3-hydroxy-3-trifluoromethyl-1-benzofuran-2 3h-one, 5,7-bis 1,1-dimethylethyl-3-hydroxy-3 trifluoromethyl-2 3h-benzofuranone
CC(C)(C)C1=CC(=C2C(=C1)C(C(=O)O2)(C(F)(F)F)O)C(C)(C)C
330.347
330.34
rac BHFF
C17H21F3O3
RVNOANDLZIIFHB-UHFFFAOYSA-N
5,7-ditert-butyl-3-hydroxy-3-(trifluoromethyl)-1-benzofuran-2-one
4332683
>98%
Videos
Documents