missing translation for 'onlineSavingsMsg'
Learn More

rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, +96%, ACROS Organics™

Brand:  Acros Organics 392230010

Code : 29

Additional Details : CAS Number : 153305-67-0 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10473195

  • £53.20 / 1g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

rac-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
≥96%
C48H40P2
MFCD01311709
153305-67-0
96+%
678.79

Specifications

rac-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
Authentic
Crystalline Powder
96+%
678.79
White to off-white
1g
153305-67-0
Glass bottle
≥96%
C48H40P2
MFCD01311709
267.0° to 270.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS