Learn More
USP10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of USP10.
Brand: Abnova PAB19258.100ug
Additional Details : Weight : 0.02000kg
Description
Ubiquitin is a highly conserved protein that is covalently linked to other proteins to regulate their function and degradation. This gene encodes a member of the ubiquitin-specific protease family of cysteine proteases. The enzyme specifically cleaves ubiquitin from ubiquitin-conjugated protein substrates. The protein is found in the nucleus and cytoplasm. It functions as a co-factor of the DNA-bound androgen receptor complex, and is inhibited by a protein in the Ras-GTPase pathway. The human genome contains several pseudogenes similar to this gene. [provided by RefSeq
Specifications
USP10 | |
Polyclonal | |
Unconjugated | |
Western Blot (1-2 ug/mL) Immunocytochemistry (20 ug/mL) The optimal working dilution should be determined by the end user. | |
USP10 | |
USP10 | |
A synthetic peptide corresponding to 16 amino acids near N-terminus of human USP10. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody rasied against synthetic peptide of USP10. | |
In PBS (0.02% sodium azide) | |
KIAA0190/MGC2621/UBPO | |
Rabbit | |
Peptide affinity purification | |
RUO | |
9100 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only