Learn More
SGMS2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SGMS2.
Brand:  Abnova PAB10380.100ug
Additional Details : Weight : 0.02000kg
Product Code. 16029236
  • £473.00 / 100µg
Estimated Shipment
09-06-2020
Specifications
Specifications
SGMS2
Unconjugated
Western blot (5 to 10 ug/mL) The optimal working dilution should be determined by the end user.
SGMS2
MGC26963|SMS2
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of SGMS2.
In PBS, pH 7.4
Liquid
SGMS2
A synthetic peptide corresponding to human SGMS2.
RUO
Primary
166929