missing translation for 'onlineSavingsMsg'
Learn More

IFNAR1 (phospho S535/S539), Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of IFNAR1.

Brand:  Abnova PAB9607.100uL

Additional Details : Weight : 0.02000kg

Product Code. 16061768

  • £459.00 / 100µL
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IFNAR1
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of IFNAR1.
In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
AVP/IFN-alpha-REC/IFNAR/IFNBR/IFRC
Rabbit
Affinity purification
RUO
3454
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Immunohistochemistry (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
IFNAR1
IFNAR1
Synthetic phosphopeptide corresponding to residues surrounding S535/S539 of human IFNAR1.
100 μL
Primary
Bovine, Canine, Human, Mouse, Primate, Rat, Sheep
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only