missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562979
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RSANALILTY DITCEESFRC LPEWLREIEQ YASNKVITVL VG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
RAB30 Polyclonal antibody specifically detects RAB30 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
RAB30 | |
Polyclonal | |
Unconjugated | |
RAB30 | |
5033421K01Rik; AI323892; EGK_06727; Rab30; RAB30, member RAS oncogene family; ras-related protein Rab-30; Rsb30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15771 | |
RAB30 | |
Recombinant protein corresponding to Human RAB30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only