Learn More
R3HCC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554710
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ENDFVHRIQE ELDRFLLQKQ LSKVLLFPPL SSRLRYLIHR TAENFDLLSS FSVGEGWKRR TVICHQDIRV PSSDGLSGPC RAPASCPSRY HGPRPISNQG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
R3HCC1 Polyclonal antibody specifically detects R3HCC1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
R3HCC1 | |
Polyclonal | |
Unconjugated | |
R3HCC1 | |
1700020M16Rik; AA407796; R3H and coiled-coil domain-containing protein 1; R3H domain and coiled-coil containing 1; R3HCC1; RGD1311790 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
203069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3T6 | |
R3HCC1 | |
Recombinant protein corresponding to Human R3HCC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only