Learn More
Saint-Gobain R3603 Vacuum Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon R3603 Pump Tubing handles virtually all inorganic chemicals found in the lab.
Brand: Saint-Gobain ACF1S1505-C
Code : 02
Additional Details : Weight : 1.00000kg
Description
Crystal clear to allow viewing for possible pump oil back-up which could cause pump damage. Extra heavy walls will withstand full vacuum of 759mmHg at 23°C and 686mmHg at 60°C. Will not swell closed should vacuum pump oil back up. Low vapour pressure makes it suitable for vacuum applications down to 10mmHg to 17mmHg at 21°C. Ideal for analytical testing of gases and vapours, it is also well suited for use in making manometers.Specifications
Tygon | |
1.5 mm | |
759 mm Hg at 80°C |
15 m | |
1 Pack |