missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-tert-Butylsulfinamide, 98%, Thermo Scientific Chemicals

Catalog Number p-4228484
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
196929-78-9
C4H11NOS
121.20
MFCD05861479
CESUXLKAADQNTB-UHFFFAOYNA-N
r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide
10964479
2-methylpropane-2-sulfinamide
CC(C)(C)S(N)=O
This item is not returnable. View return policy
196929-78-9
C4H11NOS
121.20
MFCD05861479
CESUXLKAADQNTB-UHFFFAOYNA-N
r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide
10964479
2-methylpropane-2-sulfinamide
CC(C)(C)S(N)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 99.0°C to 102.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSONH2
Quantity 1 g
Merck Index 15,3607
Specific Rotation 3.0° to 5.0° (20°C, 589nm) (c=1, chloroform)
Formula Weight 121.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-(+)-tert-Butylsulfinamide
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-tert-Butylsulfinamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'