missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R-Spondin 3 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | R-Spondin 3 |
---|---|
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human R-Spondin 3 Gln22-His272 Accession # Q9BXY4 |
Gene ID (Entrez) | 84870 |
Gene Accession No. | Q9BXY4 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
R-Spondin 3 Polyclonal specifically detects R-Spondin 3 in Human samples. It is validated for Neutralization.Specifications
R-Spondin 3 | |
84870 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human R-Spondin 3 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse R-Spondin 3 is observed. | |
Unconjugated | |
Neutralization | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Chinese hamster ovary cell line CHO-derived recombinant human R-Spondin 3 Gln22-His272 Accession # Q9BXY4 | |
Q9BXY4 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Sheep | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only