missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 1 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody has been used in 1 publication

POA

Specifications

Antigen R-Spondin 1
Immunogen Chinese hamster ovary cell line CHO-derived human R-Spondin 1 Arg31-Ala263 Accession # Q2MKA7
Gene ID (Entrez) 284654
Gene Accession No. Q2MKA7
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
View More Specs

Description

Description

R-Spondin 1 Polyclonal specifically detects R-Spondin 1 in Human samples. It is validated for Neutralization.
Specifications

Specifications

R-Spondin 1
284654
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects human R-Spondin 1 in direct ELISAs. In direct ELISAs, approximately 6% cross-reactivity with recombinant mouse R-Spondin 1 is observed and less than 1% cross-reactivity with recombinant human (rh) R-Spondin 2, rhR-Spondin 3, and rhR-Spondin 4 is observed.
Unconjugated
Neutralization
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Chinese hamster ovary cell line CHO-derived human R-Spondin 1 Arg31-Ala263 Accession # Q2MKA7
Q2MKA7
Sterile PBS to a final concentration of 0.2 mg/mL.
Sheep
IgG
Affinity Purified
RUO
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only