missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ R Series Refrigerated Baths

Brand:  Grant Instruments™ TC120-R5

Code : 57

Additional Details : Weight : 49.80000kg

Product Code. 12757819

  • £5391.75 / Each


Estimated Shipment
06-04-2018
Add to basket

Description

Description

Bath circulator system Optima TC120-R5, 12L capacity, 230mm x 115mm working area (l x w) -20°C to 100°CA powerful and efficient refrigerated circulating bath, the T100-R5 has a 12 liter tank for accurate heating or cooling in the temperature range of -20 to 100°C. Fitted with a drain tap, refrigeration high pressure switch and relay control (refrigeration on/off). Cooling power 1050W at 0°C. 230V 50/60Hz
Specifications

Specifications

12L
260 x 115mm
47kg
Videos
Documentation