missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-sec-Butanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3779943
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
14898-79-4
C4H10O
74.123
MFCD00064280
BTANRVKWQNVYAZ-SCSAIBSYSA-N
r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
84682
CHEBI:35686
(2R)-butan-2-ol
CCC(C)O
This item is not returnable. View return policy
14898-79-4
C4H10O
74.123
MFCD00064280
BTANRVKWQNVYAZ-SCSAIBSYSA-N
r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
84682
CHEBI:35686
(2R)-butan-2-ol
CCC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
Color Colorless
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula C2H5CH(OH)CH3
Beilstein 01, I, 189
Merck Index 15, 1543
Formula Weight 74.12
Percent Purity ≥98.5%
Chemical Name or Material (R)-(-)-sec-Butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (R)-(-)-sec-Butanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'