missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Rolipram, Tocris Bioscience™

PDE4 inhibitor. More active enantiomer of rolipram (Cat. No. 0905)

Brand:  Tocris (Bio-Techne) 1349/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13469203

  • £204.33 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

More active enantiomer of the PDE4 inhibitor rolipram; 2-10-fold more potent than the S-(+) enantiomer.

Available as part of the Phosphodiesterase Inhibitor Tocriset™ . Racemate and S-enantiomer also available.

Specifications

Chemical Identifiers

85416-75-7
275.348
r---rolipram, r-rolipram, r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, --rolipram, 4r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, unii-dpx51kup08, dpx51kup08, 1xmy, rolipram, r
CHEBI:40133
COC1=C(C=C(C=C1)C2CC(=O)NC2)OC3CCCC3
C16H21NO3
HJORMJIFDVBMOB-LBPRGKRZSA-N
448055
(4R)-4-(3-cyclopentyloxy-4-methoxyphenyl)pyrrolidin-2-one

Specifications

10mg
85416-75-7
r---rolipram, r-rolipram, r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, --rolipram, 4r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, unii-dpx51kup08, dpx51kup08, 1xmy, rolipram, r
COC1=C(C=C(C=C1)C2CC(=O)NC2)OC3CCCC3
275.348
CHEBI:40133
>98%
(R)-(-)-Rolipram
C16H21NO3
HJORMJIFDVBMOB-LBPRGKRZSA-N
(4R)-4-(3-cyclopentyloxy-4-methoxyphenyl)pyrrolidin-2-one
448055
275.35
Videos
Documents