missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Propylene oxide, 98+%, Thermo Scientific Chemicals

Catalog Number p-4089820
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15448-47-2
C3H6O
58.08
MFCD00066211
GOOHAUXETOMSMM-GSVOUGTGSA-N
r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
146261
CHEBI:28985
(2R)-2-methyloxirane
CC1CO1
This item is not returnable. View return policy
15448-47-2
C3H6O
58.08
MFCD00066211
GOOHAUXETOMSMM-GSVOUGTGSA-N
r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
146261
CHEBI:28985
(2R)-2-methyloxirane
CC1CO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -112.0°C
Density 0.8290g/mL
Boiling Point 33.0°C to 34.0°C
Flash Point −37°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.829
Solubility Information Solubility in water: 405g/L (20°C)
Viscosity 0.38 mPa.s (10°C)
Formula Weight 58.08
Percent Purity 98+%
Chemical Name or Material (R)-(+)-Propylene oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (R)-(+)-Propylene oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H224-Extremely flammable liquid and vapour.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'