missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals

Catalog Number p-4258865
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
616867-28-8, 1634-04-4
C21H23NO4
353.42
MFCD12031696
AWEZXIRZNQCCNN-OAQYLSRUSA-N
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
11210384
CC(C)C(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
This item is not returnable. View return policy
616867-28-8, 1634-04-4
C21H23NO4
353.42
MFCD12031696
AWEZXIRZNQCCNN-OAQYLSRUSA-N
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl, r-n-fmoc-alpha-methylvaline, fmoc-r-2-amino-2,3-dimethylbutanoicacid, fmoc-r-2-amino-2,3-dimethylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline, r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
11210384
CC(C)C(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 53°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Specific Rotation -8.6
Residual Solvents 10% max. (MTBE)
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2,3-dimethylbutanoic acid
Formula Weight 353.43
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-FMOC-α-Methylvaline, 98% ee
Show More Show Less
compliance-icons
Product Identifier
  • (R)-N-FMOC-alpha-Methylvaline
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'