missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, Thermo Scientific™

Catalog Number 11925561
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
193086-74-7
C25H22FNO4
419.45
MFCD17019319
VNYGHKCAXQKZEQ-RUZDIDTESA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine, fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine
44598473
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
CC(CC1=C(F)C=CC=C1)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
193086-74-7
C25H22FNO4
419.45
MFCD17019319
VNYGHKCAXQKZEQ-RUZDIDTESA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine, fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine
44598473
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
CC(CC1=C(F)C=CC=C1)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 122.0°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Specific Rotation 48
Formula Weight 419.46
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98% ee
Show More Show Less
Product Identifier
  • (R)-N-FMOC-alpha-Methyl-2-fluorophenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'