missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, Thermo Scientific™
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 111.0°C to 122.0°C |
Color | White to Yellow |
Enantiomeric Excess | 98.0% min. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 100 mg |
Specific Rotation | 48 |
Show More |
Safety and Handling
Product Identifier
- (R)-N-FMOC-alpha-Methyl-2-fluorophenylalanine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'