missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-α-Ethylalanine, 98%, 98% ee, Thermo Scientific Chemicals

Catalog Number p-4258770
23.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
123254-58-0
C10H19NO4
217.27
MFCD12963786
SHZXLTCEPXVCSV-SNVBAGLBSA-N
boc-d-isovaline, r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, r-2-tert-butoxycarbonylamino-2-methylbutanoic acid, boc-iso-valine, r-n-boc-a-ethylalanine, s-n-boc-a-ethylalanine, r-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2r-2-tert-butoxycarbonylamino-2-methyl-butanoic acid, 2r-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
14284791
(2R)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CCC(C)(NC(=O)OC(C)(C)C)C(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
123254-58-0
C10H19NO4
217.27
MFCD12963786
SHZXLTCEPXVCSV-SNVBAGLBSA-N
boc-d-isovaline, r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid, r-2-tert-butoxycarbonylamino-2-methylbutanoic acid, boc-iso-valine, r-n-boc-a-ethylalanine, s-n-boc-a-ethylalanine, r-2-methyl-2-tert-butoxycarbonylamino butyric acid, 2r-2-tert-butoxycarbonylamino-2-methyl-butanoic acid, 2r-2-tert-butoxy carbonyl amino-2-methylbutanoic acid, 2r-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
14284791
(2R)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CCC(C)(NC(=O)OC(C)(C)C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 104°C to 112°C
CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation -12
Formula Weight 217.27
Percent Purity 98%,98% ee
Quantity 100 mg
Physical Form Powder
Chemical Name or Material (R)-N-BOC-α-Ethylalanine, 98% ee
Show More Show Less
Product Identifier
  • (R)-N-BOC-alpha-Ethylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'