missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-Allylglycine, 95%, 98% ee, Thermo Scientific Chemicals

Catalog Number 10500362
69.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
170899-08-8
C10H17NO4
215.25
MFCD01320882
BUPDPLXLAKNJMI-SSDOTTSWSA-N
boc-d-allylglycine, r-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-alpha-allyl-d-gly, boc-d-allyglycine.dcha, 2r-2-tert-butoxycarbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, boc-d-allylglycine.dcha, d-boc-allylglycine, boc-d-allyglycine?dcha, boc-d-allyglycine?cha
638723
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
CC(C)(C)OC(=O)NC(CC=C)C(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
170899-08-8
C10H17NO4
215.25
MFCD01320882
BUPDPLXLAKNJMI-SSDOTTSWSA-N
boc-d-allylglycine, r-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-alpha-allyl-d-gly, boc-d-allyglycine.dcha, 2r-2-tert-butoxycarbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, boc-d-allylglycine.dcha, d-boc-allylglycine, boc-d-allyglycine?dcha, boc-d-allyglycine?cha
638723
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
CC(C)(C)OC(=O)NC(CC=C)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -9.5° to -12.5° (20°C, 589nm) (c=1, methanol)
Formula Weight 215.25
Percent Purity 95%,98% ee
Physical Form Viscous Liquid
Chemical Name or Material (R)-N-BOC-Allylglycine, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'