missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™

Brand:  Acros Organics 362870010

Code : 29

Additional Details : CAS Number : 261380-20-5 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10712833

  • £135.10 / 1g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

(R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
261380-20-5
100.0
95%
344.2
(R)-N-BOC-β-(4-bromophenyl)-β-alanine, (R)-N-tert-Butoxycarbonyl-3-amino-3-(4-bromophenyl)propanoic acid
95%, (98% e.e.)
94.0
≥94%
C14H18BrNO4
MFCD01320860

Specifications

(R)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid
261380-20-5
100.0
Authentic
Powder
95%
344.2
1g
(R)-N-BOC-β-(4-bromophenyl)-β-alanine, (R)-N-tert-Butoxycarbonyl-3-amino-3-(4-bromophenyl)propanoic acid
95%, (98% e.e.)
94.0
97.5% min.
Glass bottle
≥94%
C14H18BrNO4
MFCD01320860
White
Videos
SDS