missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, Thermo Scientific™

Catalog Number 10642912
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
80548-31-8
C10H15NO
165.23
MFCD01862171
GXIWMXAAPLZOBY-SECBINFHSA-N
r-+-2-1-phenylethyl amino ethanol, 2-1r-1-phenylethyl amino ethan-1-ol, 2-1r-1-phenylethyl amino ethanol, 2-r-alpha-methylbenzylamino ethanol, ethanol, 2-1r-1-phenylethyl amino, r-+-n-2-hydroxyethyl-phenylethylamine, r-+-n-2-hydroxyethyl-alpha-methylbenzylamine, r-+-n-2-hydroxyethyl-alpha-phenylethylamine
6993821
2-[[(1R)-1-phenylethyl]amino]ethanol
CC(C1=CC=CC=C1)NCCO
This item is not returnable. View return policy
80548-31-8
C10H15NO
165.23
MFCD01862171
GXIWMXAAPLZOBY-SECBINFHSA-N
r-+-2-1-phenylethyl amino ethanol, 2-1r-1-phenylethyl amino ethan-1-ol, 2-1r-1-phenylethyl amino ethanol, 2-r-alpha-methylbenzylamino ethanol, ethanol, 2-1r-1-phenylethyl amino, r-+-n-2-hydroxyethyl-phenylethylamine, r-+-n-2-hydroxyethyl-alpha-methylbenzylamine, r-+-n-2-hydroxyethyl-alpha-phenylethylamine
6993821
2-[[(1R)-1-phenylethyl]amino]ethanol
CC(C1=CC=CC=C1)NCCO
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5350
Quantity 10 g
Specific Rotation Condition +44° (22°C c=1,C2H5OH)
Specific Rotation + 44.00
Formula Weight 165.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-N-(2-Hydroxyethyl)-a-phenylethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'