missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Wnt-2b Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Accession Number | AAC25397 |
---|---|
Gene ID (Entrez) | 22414 |
Molecular Weight (g/mol) | 38kDa |
Name | Wnt-2b |
Quality Control Testing | SDS-PAGE: 41-60kDa, reducing conditions |
Description
The Recombinant Mouse Wnt-2b Protein is derived from CHO. The Recombinant Mouse Wnt-2b Protein has been validated for the following applications: Bioactivity.Specifications
AAC25397 | |
38kDa | |
SDS-PAGE: 41-60kDa, reducing conditions | |
RUO | |
Ser57 & Asp71 | |
Yes |
22414 | |
Wnt-2b | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated |