missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-Morpholine-3-carboxylic acid hydrochloride, 95%

Brand:  Alfa Aesar™ H64189.03

Code : NEW

Additional Details : CAS Number : 106825-81-4 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15488756

  • £522.58 / 1g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

(R)-Morpholine-3-carboxylic acid hydrochloride
167.59
UN2811
106825-81-4
MFCD06809588

Specifications

(R)-Morpholine-3-carboxylic acid hydrochloride
Powder
167.59
UN2811
1g
106825-81-4
95%
MFCD06809588
White
Videos
SDS