Learn More
(R)-(-)-α-Methylhistamine dihydrobromide, Tocris Bioscience™
Potent, standard H3 agonist
Brand:  Tocris (Bio-Techne) 056910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487763
  • £127.00 / 10mg
Estimated Shipment
08-03-2018
Add to basket
Description
Description
Very potent, high affinity H3 agonist (KD = 50.3 nM) that displays > 200-fold selectivity over H4 receptors. Inhibits H3-mediated histamine synthesis and release in the CNS and stimulates H4-mediated eosinophil shape change (EC50 = 66 nM). Part of the Histamine H3 Receptor Tocriset™ . (S)-(+)-α-Methylhistamine dihydrobromide (Cat. No. 0572) also available.
Specifications
Chemical Identifiers
(R)-(-)-α-Methylhistamine dihydrobromide
C6H11N3·2HBr
287
(R)-(-)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
Specifications
(R)-(-)-α-Methylhistamine dihydrobromide
868698-49-1
C6H11N3·2HBr
(R)-(-)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
10mg
>99%
287
Videos