missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride, 99%, ACROS Organics™

Brand:  Acros Organics 264120010

Code : 29

Additional Details : CAS Number : 39637-99-5 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 1a Pack group : II

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10711141

  • £240.90 / 1g


EU Stock 6
Add to basket

Specifications

Chemical Identifiers

(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride
98.5
99%
C10H8ClF3O2
MFCD00044400
39637-99-5
100.0
99%
252.62
(-)-Mosher's chloride

Specifications

(R)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride
98.5
Glass bottle
1.4680 to 1.4700
− 132.00
99%
1.3400g/mL
−132.00 (20.00°C c=6,CHCl3)
MFCD00044400
Clear colorless to light yellow
1g
(-)-Mosher's chloride
39637-99-5
100.0
Authentic
1.34
Liquid
99%
C10H8ClF3O2
252.62
213.0° to 214.0°C
15, 6365
89°C
Videos
SDS