missing translation for 'onlineSavingsMsg'
Learn More

(R)-Glycidyl butyrate, 92-94%, ACROS Organics™

Brand:  Acros Organics 252830050

Code : 29

Additional Details : CAS Number : 60456-26-0 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10003310

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(R)-Glycidyl butyrate
60456-26-0
100.0
92-94%
144.17
92-94%
92.0
≥92%
C7H12O3
MFCD00075120

Specifications

(R)-Glycidyl butyrate
60456-26-0
100.0
Liquid
92-94%
C7H12O3
MFCD00075120
-29° to -32° (20°C, 589nm) (neat)
− 30.00 (20.00°C c=neat)
5g
92-94%
92.0
Authentic
≥92%
Glass bottle
144.17
1.4270 to 1.4290 (20°C, 589nm)
1.0350g/mL
Clear colorless to slightly yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (R)-Glycidyl butyrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS