Learn More
(R)-(+)-Etomoxir sodium salt, Tocris Bioscience™
Carnitine palmitoyltransferase I (CPT1) inhibitor
Brand:  RD Systems (Bio-Techne) 4539/10
Code : Z1
Additional Details : Weight : 0.00010kg
Quantity 10mg
 View more versions of this product
Product Code. 13454904
  • £189.63 / 10mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Inhibitor of carnitine palmitoyltransferase I (CPT1); inhibits β-oxidation in mitochondria. Shown to inhibit cardiolipin biosynthesis from exogenous fatty acid in H9c2 cells.
Specifications
Chemical Identifiers
828934-41-4
321.753
(R)-(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt
(2R)-2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylic acid;sodium
C15H19ClNaO4
ZUMIFLJZQDMWAU-XFULWGLBSA-N
131860301
C1C(O1)(CCCCCCOC2=CC=C(C=C2)Cl)C(=O)O.[Na]
Specifications
10mg
828934-41-4
(R)-(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt
C1C(O1)(CCCCCCOC2=CC=C(C=C2)Cl)C(=O)O.[Na]
321.753
320.74
(R)-(+)-Etomoxir sodium salt
C15H19ClNaO4
ZUMIFLJZQDMWAU-XFULWGLBSA-N
(2R)-2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylic acid;sodium
131860301
>98%