Learn More
(R)-(+)-Etomoxir sodium salt, Tocris Bioscience™
Carnitine palmitoyltransferase I (CPT1) inhibitor
POA
Description
Description
Inhibitor of carnitine palmitoyltransferase I (CPT1); inhibits β-oxidation in mitochondria. Shown to inhibit cardiolipin biosynthesis from exogenous fatty acid in H9c2 cells.