Learn More
(R)-(+)-Etomoxir sodium salt, Tocris Bioscience™
Carnitine palmitoyltransferase I (CPT1) inhibitor
Brand:  RD Systems (Bio-Techne) 4539/50
Code : Z1
Additional Details : Weight : 0.00050kg
Quantity 50mg
 View more versions of this product
Product Code. 13464904
  • £600.29 / 50mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Inhibitor of carnitine palmitoyltransferase I (CPT1); inhibits β-oxidation in mitochondria. Shown to inhibit cardiolipin biosynthesis from exogenous fatty acid in H9c2 cells.
Specifications
Chemical Identifiers
828934-41-4
321.753
(R)-(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt
(2R)-2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylic acid;sodium
C15H19ClNaO4
ZUMIFLJZQDMWAU-XFULWGLBSA-N
131860301
C1C(O1)(CCCCCCOC2=CC=C(C=C2)Cl)C(=O)O.[Na]
Specifications
(R)-(+)-Etomoxir sodium salt
828934-41-4
(R)-(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt
C1C(O1)(CCCCCCOC2=CC=C(C=C2)Cl)C(=O)O.[Na]
321.753
320.74
50mg
C15H19ClNaO4
ZUMIFLJZQDMWAU-XFULWGLBSA-N
(2R)-2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylic acid;sodium
131860301
>98%
Videos