missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Viral Variola CRMB Recombinant Protein

Brand:  R&D Systems™ 4778-CR-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465074

  • £352.00 / 10µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala23-Leu349, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.5 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS
Variola CRMB
10μg
Disulfide-linked homodimer
Yes
36.5 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Viral
Unlabeled
Videos