missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Viral MIP-I Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Molecular Weight (g/mol) | 7.9 kDa |
---|---|
Name | MIP-I |
Quantity | 25μg |
Source | E. coli-derived viral MIP-I protein Ala25-Ala95 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Description
The Recombinant Viral MIP-I Protein is derived from E. coli. The Recombinant Viral MIP-I Protein has been validated for the following applications: Bioactivity.Specifications
7.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
MIP-I | |
E. coli-derived viral MIP-I protein Ala25-Ala95 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |