Learn More
R&D Systems™ TACS-XL In Situ Apoptosis Detection Kits
A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy
Brand:  R&D Systems™ 4828-30-R
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13445124
  • £336.61 / 30 tests
Estimated Shipment
01-07-2019
Includes: Proteinase K Solution; Cytonin; TdT enzyme, B-dNTP mix; Strep-HRP; anti-BrdU; Strep-Diluent
Description
Description
Based on incorporation of bromodeoxyuridine (BrdU) at the 3 ft. OH ends of the DNA fragments that form during apoptosis
This detection system uses a biotin conjugated anti-BrdU antibody and streptavidin-horseradish peroxidase for visualiza
Specifications
Specifications
30 Tests
TACS-XL In Situ Apoptosis Detection Kit
Replenisher
Videos