missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TACS-XL In Situ Apoptosis Detection Kits

A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy

Brand:  R&D Systems™ 4828-30-DK

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13435124

  • £498.33 / 30 tests

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Includes: Proteinase K Solution; Cytonin; TdT enzyme, B-dNTP mix; Strep-HRP; anti-BrdU; Strep-Diluent

Description

Description

Based on incorporation of bromodeoxyuridine (BrdU) at the 3 ft. OH ends of the DNA fragments that form during apoptosis
This detection system uses a biotin conjugated anti-BrdU antibody and streptavidin-horseradish peroxidase for visualiza

Specifications

Specifications

30 Tests
TACS-XL In Situ Apoptosis Detection Kit
DAB
The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Videos
Documents